Learn More
C6orf150 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 26416-1-AP-150UL
 View more versions of this product
Product Code. 16874885
  • £349.00 / 150µL
Description
The exact function of C6orf150 remains unknown.
Specifications
Specifications
C6orf150
Polyclonal
Unconjugated
CGAS
C6orf150, cGAS, Cyclic GMP-AMP synthase, h-cGAS, MB21D1
Rabbit
Antigen Affinity Chromatography
RUO
115004
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Western Blot
0.23 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8N884
CGAS
C6orf150 Fusion Protein Ag23455
150 μL