missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ C6orf106 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29022
This item is not returnable.
View return policy
Description
C6orf106 Polyclonal antibody specifically detects C6orf106 in Human,Mouse samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| C6orf106 | |
| Polyclonal | |
| Tris-citrate/phosphate buffer, pH 7 to 8 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 64771 | |
| Store at 4°C. Do not freeze. | |
| IgG |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| chromosome 6 open reading frame 106, dJ391O22.4, FLJ22195, FLJ32402, FP852, MGC4614, RP3-391O22.4 | |
| Between 248 and 298 | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction