missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C6orf10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5140313
This item is not returnable.
View return policy
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: ILTGYMDEEL AKKSCSKIQI LKCGGTARSQ NSREENKEAL KNDIIFTNSV ESLKSAHIKE PEREGKGTDL EKDKIGMEVK VDSDAGIP.
The function of this protein remains unknown.
Specifications
| C6orf10 | |
| Polyclonal | |
| Unconjugated | |
| TSBP1 | |
| C6orf10; chromosome 6 open reading frame 10; testis specific basic protein; Testis-expressed basic protein 1; TSBP; TSBP1; Uncharacterized protein C6orf10 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10665 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5SRN2 | |
| TSBP1 | |
| Recombinant protein corresponding to Human TSBP1. Recombinant protein control fragment (Product #RP-108860). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction