missing translation for 'onlineSavingsMsg'
Learn More

C646, MedChemExpress

Product Code. 16495878
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16495878 10 mg 10mg
16492027 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16495878 Supplier Medchem Express Supplier No. HY13823
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress C646 is a selective and competitive histone acetyltransferase p300 inhibitor with Ki of 400 nM, and is less potent for other acetyltransferases.

C646 is a selective and competitive histone acetyltransferase p300 inhibitor with Ki of 400 nM, and is less potent for other acetyltransferases.

Specifications

Chemical Name or Material C646
CAS 328968-36-1
Color Red Brown
Purity Grade Notes Research
Molecular Formula C24H19N3O6
Quantity 10 mg
Solubility Information DMSO : 16.67 mg/mL (37.43 mM; Need ultrasonic)
SMILES O=C(O)C1=CC=C(N2N=C(C)/C(C2=O)=C\C3=CC=C(C4=CC(C)=C(C)C=C4[N+]([O-])=O)O3)C=C1
Molecular Weight (g/mol) 445.42
Formula Weight 445.42
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.