missing translation for 'onlineSavingsMsg'
Learn More

C6 Ceramide, TRC

Product Code. 30363108
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30363108 25 mg 25mg
30355143 50 mg -
30363007 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30363108 Supplier Toronto Research Chemicals Supplier No. TRCC26260025MG
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124753-97-5
Molecular Formula C24 H47 N O3
Molecular Weight (g/mol) 397.63
Synonym N-Hexanoyl-D-erythro-Sphingosine
IUPAC Name N-[(E,2R,3S)-1,3-dihydroxyoctadec-4-en-2-yl]hexanamide
SMILES CCCCCCCCCCCCC\C=C\[C@H](O)[C@@H](CO)NC(=O)CCCCC

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C24H47NO3/c1-3-5-7-8-9-10-11-12-13-14-15-16-18-19-23(27)22(21-26)25-24(28)20-17-6-4-2/h18-19,22-23,26-27H,3-17,20-21H2,1-2H3,(H,25,28)/b19-18+/t22-,23+/m1/s1
Formula Weight 397.3556
Chemical Name or Material C6 Ceramide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.