missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
C5orf60 Polyclonal antibody specifically detects C5orf60 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Formulation | 50mM Sodium Borate |
| Gene Alias | Chromosome 5 Open Reading Frame 60, Putative Uncharacterized Protein C5orf60, Putative Uncharacterized Protein FLJ35723 |
| Host Species | Rabbit |
| Gene ID (Entrez) | 285679 |
| Content And Storage | Store at 4C in the dark. |
| Form | Purified |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?