missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C5orf51 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560506
This item is not returnable.
View return policy
Description
Immunogen sequence: IFSDIHLLAM MYSGEMCYWG SKYCADQQPE NHEVDTSVSG AGCTTYKEPL DFREVGEKIL KKYVSVCEGP LKEQEWNTTN AKQILNFFHH RCN Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 23%.
C5orf51 is a protein coding gene.
Specifications
| C5orf51 | |
| Polyclonal | |
| Unconjugated | |
| C5orf51 | |
| C5orf51; chromosome 5 open reading frame 51; UPF0600 protein; UPF0600 protein C5orf51 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 285636 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| A6NDU8 | |
| C5orf51 | |
| Recombinant protein corresponding to Human C5orf51. Recombinant protein control fragment (Product #RP-100350). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction