missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C5orf33 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26352-1-AP-20UL
This item is not returnable.
View return policy
Description
Mitochondrial NAD(+) kinase that phosphorylates NAD(+) to yield NADP(+). Can use both ATP or inorganic polyphosphate as the phosphoryl donor. Also has weak NADH kinase activity in vitro; however NADH kinase activity is much weaker than the NAD(+) kinase activity and may not be relevant in vivo.Specifications
| C5orf33 | |
| Polyclonal | |
| Unconjugated | |
| NADK2 | |
| C5orf33, NAD kinase 2, NADK2, UPF0465 protein C5orf33 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 133686, 365699, 68646 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot | |
| 0.37 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q1HCL7, Q4G0N4, Q8C5H8 | |
| Nadk2 | |
| C5orf33 Fusion Protein Ag23706 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction