missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C5orf24 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | C5orf24 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
C5orf24 Polyclonal specifically detects C5orf24 in Human samples. It is validated for Western Blot.Specifications
| C5orf24 | |
| Polyclonal | |
| Rabbit | |
| Q7Z6I8 | |
| 134553 | |
| Synthetic peptides corresponding to C5ORF24 The peptide sequence was selected from the middle region of C5orf24. Peptide sequence SIEIKDELKKKKNLNRSGKRGRPSGTTKSAGYRTSTGRPLGTTKAAGFKT. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| chromosome 5 open reading frame 24, FLJ37562, hypothetical protein LOC134553 | |
| C5ORF24 | |
| IgG | |
| 20 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title