missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C5b-9 Antibody (2A1), Novus Biologicals™
Mouse Monoclonal Antibody
£137.00 - £329.00
Specifications
| Antigen | C5b-9 |
|---|---|
| Clone | 2A1 |
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoassay |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18397710
|
Novus Biologicals
NBP2-77377-0.025MG |
0.025 mg |
£137.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18769793
|
Novus Biologicals
NBP2-77377-0.1MG |
0.1 mg |
£329.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
C5b-9 Monoclonal specifically detects C5b-9 in Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoassay.Spécification
| C5b-9 | |
| 0.5 mg/mL | |
| Monoclonal | |
| Purified | |
| PBS | |
| Rat C5b-9 | |
| Store at 4°C. |
| 2A1 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoassay | |
| Unconjugated | |
| Mouse | |
| MAC, Membrane attack complex, sC5-b9, sC5b-9, TCC, Terminal complement complex | |
| IgG1 |
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit