missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C4orf47 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560456
This item is not returnable.
View return policy
Description
Immunogen sequence: ITVGDKYVSQ FNRPFNEAAS KNKQMLPGGS KEMSDLQAGY FDPHFVRIFE GEGYINLNQV RRRDMVEAAK KNLGKAFLPS N Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 81%.
C4orf47 is a protein coding gene.
Specifications
| C4orf47 | |
| Polyclonal | |
| Unconjugated | |
| C4orf47 | |
| C4orf47; Chr4_1746; chromosome 4 open reading frame 47; UPF0602 protein C4orf47 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 441054 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| A7E2U8 | |
| C4orf47 | |
| Recombinant protein corresponding to Human C4orf47. Recombinant protein control fragment (Product #RP-100318). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction