missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C4orf34 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA562372
This item is not returnable.
View return policy
Description
Immunogen sequence: AEGGFDPCEC VCSHEHAMRR LINLLRQSQS YCTDTECL Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 97%.
Specifications
| C4orf34 | |
| Polyclonal | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96QK8 | |
| SMIM14 | |
| Recombit protein corresponding to Human C4orf34 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry | |
| Unconjugated | |
| SMIM14 | |
| uncharacterized protein C4orf34 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 201895 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Research Use Only
Spot an opportunity for improvement?Share a Content Correction