missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C4orf31 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5107005
This item is not returnable.
View return policy
Description
Sequence homology: Human: 100%; Pig: 86%; Zebrafish: 76%.
Promotes matrix assembly and cell adhesiveness. Promotes neuron migration, growth and survival as well as neurite outgrowth (PubMed:20969804). Promotes endothelial cell survival, vessel formation and plays an important role in the process of revascularization through NOS3-dependent mechanisms (PubMed:24706764). [UniProt].
Specifications
| C4orf31 | |
| Polyclonal | |
| Unconjugated | |
| NDNF | |
| C4orf31; fibronectin type-III domain-containing protein C4orf31; NDNF; neuron derived neurotrophic factor; neuron-derived neurotrophic factor; NORD; Protein NDNF; UNQ2748/PRO6487 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 79625 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 | |
| Q8TB73 | |
| NDNF | |
| Synthetic peptide directed towards the N-terminal region of Human NDNF. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction