missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C4orf3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113166
This item is not returnable.
View return policy
Description
C4orf3 Polyclonal Antibody for Western Blot, ELISA
C4orf3 is a protein coding gene.
Specifications
| C4orf3 | |
| Polyclonal | |
| Unconjugated | |
| C4orf3 | |
| C4orf3; chromosome 4 open reading frame 3; HCV F-transactivated protein 1; Hepatitis C virus F protein-transactivated protein 1; uncharacterized protein C4orf3 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 401152 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q8WVX3 | |
| C4orf3 | |
| Recombinant Human Uncharacterized protein C4orf3 protein (1-44AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction