missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C4orf14 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA557421
This item is not returnable.
View return policy
Description
Immunogen sequence: LVAEDIMLKE GLGASEAVAD IKFSSAGWVS VTPNFKDRLH LRGYTPEGTV LTVRPPLLPY IVNIKGQRIK KSVAYKTKKP PSLMYNVRKK KGKIN Highest antigen sequence identity to the following orthologs: Mouse - 72%, Rat - 72%.
The protein encoded by this gene is a nuclear-encoded GTPase that functions in the mitochondrion. Upon translation, this protein is imported into the nucleus and then into the nucleolus before being exported to the mitochondrion. The encoded protein is required for oxygen-dependent regulation of mitochondrial respiratory complexes and for mitochondrial protein synthesis.
Specifications
| C4orf14 | |
| Polyclonal | |
| Unconjugated | |
| NOA1 | |
| 2610024G14Rik; AtNOS1; AU022345; C4orf14; hAtNOS1; hNOA1; mAtNOS1; mitochondrial GTPase 3 homolog; MMR_HSR1 domain containing protein RGD1359460; MNCb-4931; MTG3; nitric oxide associated 1; nitric oxide associated-1; nitric oxide synthase, mitochondrial (putative); nitric oxide-associated protein 1; Noa1; putative ortholog of Arabidopsis mitochondrial nitric oxide synthase; RGD1359460 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84273 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8NC60 | |
| NOA1 | |
| Recombinant protein corresponding to Human C4orf14 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Research Use Only
Spot an opportunity for improvement?Share a Content Correction