missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C4BPB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA576914
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Complement component 4-binding protein (C4BP) is a plasma glycoprotein that inhibits the classical pathway of complement activation, which is mediated through antibody targeting of foreign antigen. Structurally, C4BP is a disulfide linked, multimeric protein that is composed of seven alpha chains and one beta chain. C4BP functions as a cofactor for C3beta inactivator in the cleavage of C3beta, and accelerates the decay of C4betaC2alpha (C3 convertase) by acting as a cofactor in the cleavage of C4beta by factor I. Streptococcal strains that express Ig-binding cell surface molecules, which are members of the M protein family, can bind to overlapping C4beta binding sites in C4BP and therefore, interfere with the classical pathway of complement activation. Bacteria-bound C4BP may be an evolved mechanism that downregulates complement activation in the bacterial host microenvironment, thereby reducing the occurrences of bacterial opsonization and phagocytosis.
Specifications
| C4BPB | |
| Polyclonal | |
| Unconjugated | |
| C4BPB | |
| C4b-binding protein beta chain; C4BP; C4BPB; C4bp-ps1; complement component 4 binding protein beta; complement component 4 binding protein, beta; complement component 4 binding protein, pseudogene 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 12270, 24236, 725 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| P20851, Q63515 | |
| C4bp-ps1, C4BPB | |
| Recombinant full length Human C4BPB. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction