missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C4 gamma chain Mouse anti-Human, Clone: 2G6A7, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | C4 gamma chain |
|---|---|
| Clone | 2G6A7 |
| Concentration | 0.81 mg/mL |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16867935
|
Proteintech
66242-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16857935
|
Proteintech
66242-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This antibody raised against 1543-1744aa of human C4-A protein recognizes C4 gamma chain.
This gene encodes the acidic form of complement factor 4, part of the classical activation pathway. The protein is expressed as a single chain precursor which is proteolytically cleaved into a trimer of alpha, beta, and gamma chains prior to secretion. The trimer provides a surface for interaction between the antigen-antibody complex and other complement components. The alpha chain may be cleaved to release C4 anaphylatoxin, a mediator of local inflammation. Deficiency of this protein is associated with systemic lupus erythematosus and type I diabetes mellitus. This gene localizes to the major histocompatibility complex (MHC) class III region on chromosome 6. Varying haplotypes of this gene cluster exist, such that individuals may have 1, 2, or 3 copies of this gene.Specifications
| C4 gamma chain | |
| 0.81 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Acidic complement C4, C4, C4A, C4A2, C4A3, C4A4, C4A6, C4S, CO4, Complement C4 A, CPAMD2, RG | |
| C4A | |
| IgG1 | |
| Protein G | |
| Antibody |
| 2G6A7 | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human | |
| P0C0L4 | |
| 720 | |
| C4 gamma chain Fusion Protein Ag10449 | |
| Primary | |
| -20°C | |
| C4A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title