missing translation for 'onlineSavingsMsg'
Learn More

C4 binding protein B Antibody (1G9), Novus Biologicals™

Product Code. 18354318 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18354318 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18354318 Supplier Novus Biologicals Supplier No. H00000725M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

C4 binding protein B Monoclonal antibody specifically detects C4 binding protein B in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen C4 binding protein B
Applications Western Blot, ELISA, Immunoprecipitation, Sandwich ELISA
Classification Monoclonal
Clone 1G9
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH05378
Gene Alias C4b binding protein, beta chain, C4b-binding protein beta chain, C4BP, complement component 4 binding protein, beta, complement component 4 binding protein, beta chain, complement component 4-binding protein, beta
Host Species Mouse
Immunogen C4BPB (AAH05378, 1 a.a. ~ 251 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MFFWCACCLMVAWRVSASDEHCPELPPVDNSIFVAKEVEGQILGTYVCIKGYHLVGKKTLFCNASKEWDNTTTECRLGHCPDPVLVNGEFSSSGPVNVSDKITFMCNDHYILKGSNRSQCLEDHTWAPPFPICKSRDCDPPGNPVHGYFEGNNFTLGSTISYYCEDRYYLVGVQEQQCVDGEWSSALPVCKLIQEAPKPECEKALLAFQESKNLCEAMENFMQQLKESGMTMEELKYSLELKKAELKAKLL
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 725
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.