missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C4.4A/LYPD3 Antibody, Alexa Fluor™ 700, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98298AF700
This item is not returnable.
View return policy
Description
C4.4A/LYPD3 Polyclonal specifically detects C4.4A/LYPD3 in Mouse samples. It is validated for ELISA.
Specifications
| C4.4A/LYPD3 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 27076 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| Alexa Fluor 700 | |
| C4.4A2310061G07Rik, GPI-anchored metastasis-associated protein C4.4A homolog, GPI-anchored metastasis-associated protein homolog, LY6/PLAUR domain containing 3, ly6/PLAUR domain-containing protein 3, Matrigel-induced gene C4 protein, MIG-C4 | |
| Produced in rabbits immunized with purified, recombinant C4.4A/LYPD3 (NP_598504.1; Met 1-His 287). | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction