missing translation for 'onlineSavingsMsg'
Learn More

C4.4A/LYPD3 Antibody (213), DyLight 350, Novus Biologicals™

Product Code. 18366750 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18366750 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18366750 Supplier Novus Biologicals Supplier No. NBP289995UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

C4.4A/LYPD3 Monoclonal specifically detects C4.4A/LYPD3 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C4.4A/LYPD3
Applications Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone 213
Conjugate DyLight 350
Formulation 50 mM Sodium Borate
Gene Alias C4.4A2310061G07Rik, GPI-anchored metastasis-associated protein C4.4A homolog, GPI-anchored metastasis-associated protein homolog, LY6/PLAUR domain containing 3, ly6/PLAUR domain-containing protein 3, Matrigel-induced gene C4 protein, MIG-C4
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human C4.4A/LYPD3 (NP_055215.2; Met 1-His 286).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27076
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.