missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C3orf70 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559760
This item is not returnable.
View return policy
Description
Immunogen sequence: ARRPDFQPCD GLSICATHSH GKCFKLHWCC HLGWCHCKYM YQPMTPVEQL PSTEIPARPR EPTNTIQISV SLTEHFLKFA SVFQ Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.
C3orf70 is a protein coding gene.
Specifications
| C3orf70 | |
| Polyclonal | |
| Unconjugated | |
| C3orf70 | |
| C3orf70; chromosome 3 open reading frame 70; UPF0524 protein C3orf70 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 285382 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| A6NLC5 | |
| C3orf70 | |
| Recombinant protein corresponding to Human C3orf70. Recombinant protein control fragment (Product #RP-100273). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction