missing translation for 'onlineSavingsMsg'
Learn More

C3orf38 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18349012 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
50 μg
Unit Size:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18349012 50 μg 50µL
30029110 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18349012 Supplier Novus Biologicals Supplier No. NBP31464450UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

C3orf38 Polyclonal antibody specifically detects C3orf38 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen C3orf38
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000
Formulation PBS, pH 7.0 with 0.03% Proclin 300
Gene Alias chromosome 3 open reading frame 38, FLJ54270, hypothetical protein LOC285237, MGC26717
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human C3orf38 fragment
Purification Method Antigen and protein A Affinity-purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 285237
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.