missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C3orf31 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113300
This item is not returnable.
View return policy
Description
C3orf31 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
C3orf31 may be involved in the translocation of transit peptide-containing proteins across the mitochondrial inner membrane.
Specifications
| C3orf31 | |
| Polyclonal | |
| Unconjugated | |
| TAMM41 | |
| 1500001M20Rik; C20H3orf31; C22H3orf31; C3orf31; CDP-DAG synthase; CDP-diacylglycerol synthase; Mitochondrial translocator assembly and maintenance protein 41 homolog; MMP37-like protein, mitochondrial; phosphatidate cytidylyltransferase, mitochondrial; RAM41; TAM41; TAM41 mitochondrial translocator assembly and maintenance homolog; TAM41, mitochondrial translocator assembly and maintenance protein, homolog; TAM41, mitochondrial translocator assembly and maintenance protein, homolog (S. cerevisiae); TAMM41; Unknown (protein for MGC:134188) | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 132001, 68971 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q3TUH1, Q96BW9 | |
| TAMM41 | |
| Recombinant Human Phosphatidate cytidylyltransferase, mitochondrial protein (161-452AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction