missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAPGEF1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5117396
This item is not returnable.
View return policy
Description
Centrifuge briefly prior to opening.
This gene encodes a human guanine nucleotide exchange factor. It transduces signals from CRK by binding the SH3 domain of CRK, and activating several members of the Ras family of GTPases. This signaling cascade that may be involved in apoptosis, integrin-mediated signal transduction, and cell transformation. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some variants has not been determined.
Specifications
| RAPGEF1 | |
| Polyclonal | |
| Unconjugated | |
| RAPGEF1 | |
| 4932418O06Rik; C3G; C3G-1; C3G-2; CRK SH3-binding GNRP; GRF2; guanine nucleotide releasing factor 2; Guanine nucleotide-releasing factor 2; guanine nucleotide-releasing factor 2 (specific for crk proto-oncogene); Protein C3G; Rap guanine nucleotide exchange factor (GEF) 1; Rap guanine nucleotide exchange factor 1; Rapgef1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 107746, 2889 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1.23 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| Q13905 | |
| RAPGEF1 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human C3G. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction