missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C3a Receptor Recombinant Rabbit Monoclonal Antibody (HL2744)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556615
This item is not returnable.
View return policy
Description
C3a Receptor Recombinant Monoclonal Antibody for Western Blot, ICC/IF
C3AR1 is a receptor for the chemotactic and inflammatory peptide anaphylatoxin C3a. The receptor stimulates chemotaxis, granule enzyme release and superoxide anion production. (Uniprot).
Specifications
| C3a Receptor | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q16581 | |
| C3ar1 | |
| Recombinant protein encompassing a sequence within the Extracellular domain of human C3a Receptor. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| HL2744 | |
| Unconjugated | |
| C3ar1 | |
| anaphylatoxin C3a receptor; AZ3B; C3a anaphylatoxin chemotactic receptor; C3a receptor; C3AR; C3a-R; C3AR1; C3r1; complement component 3 receptor 1; complement component 3a receptor 1; HNFAG09 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 719 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction