missing translation for 'onlineSavingsMsg'
Learn More

uvex C300 Wet Plus Safety Gloves

Product Code. 17270441 Shop All Uvex Products
Click to view available options
Size:
7
8
9
10
11
Unit Size:
10 pairs
This item is not returnable. View return policy

Product Code. 17270441

Brand: uvex 6054610

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Bamboo TwinFlex™ Technology provides level C cut protection, while SoftGrip foam coating on the palm and fingertips enhances grip capabilities.

  • SoftGrip coating on the palm and fingertips for a high level of abrasion resistance
  • Bamboo TwinFlex™ Technology provides level C cut protection
  • Climazone™ ensures the highest possible user comfort
  • Enhanced grip is wet areas and oily working conditions
  • Free of harmful substances in accordance with Oeko-Tex™ Standard 100
  • Silicone-free according to imprint test
  • Resistant to oil and grease
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance OEKO-TEX™ Standard 100
Applications Industrial
Coating Material HPE
Color Gray, Black
Hand Ambidextrous
Latex Free Yes
Material Bamboo, HPPE, Polyamide
Product Type Cut-resistant Gloves
Size 10
EN388 Standard 4X42C
For Use With (Application) Automotive, Maintenance, Metal Work, and Precision Assembly
Gender Unisex
Glove Cuff Style Knit
Series C300 Wet Plus
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.