missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C3 Goat anti-Mouse, HRP, Polyclonal , Abnova™
Goat polyclonal antibody raised against mouse C3.
Brand: Abnova PAB29686.100ug
This item is not returnable.
View return policy
Specifications
| C3 | |
| Polyclonal | |
| Goat polyclonal antibody raised against mouse C3. | |
| In 50mM HEPES (Stabilizer, 0.002% Thimerosal) | |
| C3 | |
| Native purified mouse C3. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Liquid |
| ELISA, Immunohistochemistry, Western Blot | |
| HRP | |
| ELISA (0.01-0.1 ug/mL) Immunohistochemistry (0.5-5.0 ug/mL) Western Blot (0.05-0.5 ug/mL) The optimal working dilution should be determined by the end user. | |
| AI255234/ASP/Plp | |
| Goat | |
| Antigen Affinity Purification | |
| RUO | |
| 12266 | |
| Store at 4°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction