missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C2orf83 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA556437
This item is not returnable.
View return policy
Description
Immunogen sequence: PGMRKLTVQT CGLTFIHPAG HGLCHPTAQA SAETLSSTAL NRPSVREGAC NEKSTENKKP QDSVLWS Highest antigen sequence identity to the following orthologs: Mouse - 30%, Rat - 31%.
Specifications
| C2orf83 | |
| Polyclonal | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q53S99 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 56918 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry | |
| Unconjugated | |
| C2orf83 | |
| C2ORF83 | |
| Recombit protein corresponding to Human C2orf83 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Research Use Only
Spot an opportunity for improvement?Share a Content Correction