missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C2orf57 Polyclonal Antibody
C2orf57 Polyclonal Antibody
Brand: Thermo Scientific PA563302
This item is not returnable.
View return policy
Description
C2orf57 Polyclonal Antibody for IHC
Specifications
| C2orf57 | |
| Polyclonal | |
| Unconjugated | |
| TEX44 | |
| C2orf57; chromosome 2 open reading frame 57; Testis-expressed protein 44; TEX44; uncharacterized protein C2orf57 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 165100 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q53QW1 | |
| TEX44 | |
| Recombinant protein corresponding to Human C2orf57. Recombinant protein control fragment (Product #RP-99692). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction