Learn More
Invitrogen™ C2orf47 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113817
Description
Immunogen sequence: GASVFQVKLG NQNVETKQLL SASYEFQREF TQGVKPDWTI ARIEHSKLLE For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 85%.
Promotes sorting of SMDT1/EMRE in mitochondria by ensuring its maturation. Interacts with the transit peptide region of SMDT1/EMRE precursor protein in the mitochondrial matrix, leading to protect it against protein degradation by YME1L1, thereby ensuring SMDT1/EMRE maturation by the mitochondrial processing peptidase (PMPCA and PMPCB).
Specifications
| C2orf47 | |
| Polyclonal | |
| Unconjugated | |
| MAIP1 | |
| 9430016H08Rik; AA409999; C2orf47; C78988; chromosome 2 open reading frame 47; m-AAA protease-interacting protein 1, mitochondrial; Maip1; Matrix AAA peptidase-interacting protein 1; MGC94335; RIKEN cDNA 9430016H08 gene; similar to hypothetical protein FLJ22555; uncharacterized protein C2orf47 homolog, mitochondrial; uncharacterized protein C2orf47, mitochondrial | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 79568 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8WWC4 | |
| MAIP1 | |
| Synthetic peptide directed towards the middle region of human C2orf47. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.