missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C2orf47 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566656
This item is not returnable.
View return policy
Description
Immunogen sequence: FAHVSKLLSQC KFDLLEELVA KEVLHALKEK VTSLPDNHKN ALAANIDEIV Highest antigen sequence identity to the following orthologs - mouse 88%, rat 86%.
Promotes sorting of SMDT1/EMRE in mitochondria by ensuring its maturation. Interacts with the transit peptide region of SMDT1/EMRE precursor protein in the mitochondrial matrix, leading to protect it against protein degradation by YME1L1, thereby ensuring SMDT1/EMRE maturation by the mitochondrial processing peptidase (PMPCA and PMPCB).
Specifications
| C2orf47 | |
| Polyclonal | |
| Unconjugated | |
| MAIP1 | |
| 9430016H08Rik; AA409999; C2orf47; C78988; chromosome 2 open reading frame 47; m-AAA protease-interacting protein 1, mitochondrial; Maip1; Matrix AAA peptidase-interacting protein 1; MGC94335; RIKEN cDNA 9430016H08 gene; similar to hypothetical protein FLJ22555; uncharacterized protein C2orf47 homolog, mitochondrial; uncharacterized protein C2orf47, mitochondrial | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79568 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8WWC4 | |
| MAIP1 | |
| Recombinant Human C2orf47. Recombinant protein control fragment (Product #RP-105297). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction