Learn More
C2orf3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 26029-1-AP-150UL
 View more versions of this product
Product Code. 16874555
  • £349.00 / 150µL
Description
The MW of this protein is 89 kDa, and this antibody specifically recognizes the 89 kDa protein.
TCF-9 (transcription factor 9), also known as C2orf3 (chromosome 2 open reading frame 3), GCF (GC-rich sequence DNA-binding factor) or DNABF, is a 781 amino acid nuclear protein that belongs to the GCF family. Widely expressed, GCF binds the GC-rich sequences of beta-Actin, EGFR and calcium-dependent protease (CANP) promoters. GCF contains multiple phosphoserine and phosphothreonine residues, and two GCF isoforms are produced due to alternative splicing events. GCF is considered a candidate for susceptibility to dyslexia (DYX3) as both genes reside in close proximity on human chromosome 2. Chromosome 2 is the second largest human chromosome and consists of 237 million bases, encodes over 1,400 genes and makes up approximately 8% of the human genome.
Specifications
Specifications
C2orf3
Polyclonal
Unconjugated
GCFC2
C2orf3, DNABF, GCF, TCF 9, TCF9, Transcription factor 9
Rabbit
Antigen Affinity Chromatography
RUO
6936
-20°C
Liquid
Western Blot
0.33 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P16383
GCFC2
C2orf3 Fusion Protein Ag23315
150 μL