missing translation for 'onlineSavingsMsg'
Learn More

C2GNT3 Antibody, Novus Biologicals™

Product Code. 18399659 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18399659 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18399659 Supplier Novus Biologicals Supplier No. H00051301B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

C2GNT3 Polyclonal antibody specifically detects C2GNT3 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen C2GNT3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Accession No. NP_057675.1
Gene Alias beta-1,3-galactosyl-O-glycosyl-glycoproteinbeta-1,6-N-acetylglucosaminyltransferase 4, C2GNT3, core 2 beta-1,6-N-acetylglucosaminyltransferase 3, Core 2-branching enzyme 3, Core2-GlcNAc-transferase 3, EC 2.4.1.102, glucosaminyl (N-acetyl) transferase 4, core 2, glucosaminyl (N-acetyl) transferase 4, core 2(beta-1,6-N-acetylglucosaminyltransferase)
Host Species Mouse
Immunogen GCNT4 (NP_057675.1, 1 a.a. - 453 a.a.) full-length human protein. MKIFKCYFKHTLQQKVFILFLTLWLLSLLKLLNVRRLFPQKDIYLVEYSLSTSPFVRNRYTHVKDEVRYEVNCSGIYEQEPLEIGKSLEIRRRDIIDLEDDDVVAMTSDCDIYQTLRGYAQKLVSKEEKSFPIAYSLVVHKDAIMVERLIHAIYNQHNIYCIHYDRKAPDTFKVAMNNLAKCFSNIFIASKLEAVEYAHISRLQADLNCLSDLLKSSIQWKYVINLCGQDFPLKSNFELVSELKKLNGANMLETVKPPNSKLERFTYHHELRRVPYEYVKLPIRTNISKEAPPHNIQIFVGSAYFVLSQAFVKYIFNNSIVQDFFAWSKDTYSPDEHFWATLIRVPGIPGEISRSAQDVSDLQSKTRLVKWNYYEGFFYPSCTGSHLRSVCIYGAAELRWLIKDGHWFANKFDSKVDPILIKCLAEKLEEQQRDWITLPSEKLFMDRNLTTTS
Purification Method Protein G purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51301
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.