missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C2CD5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA546763
This item is not returnable.
View return policy
Description
Peptide sequence: RAIGTACTLD KLSSPAAFLP ACNSPSKEMK EIPFNEDPNP NTHSSGPSTP.
C2CD5 gene ontology annotations related to this gene include calcium-dependent phospholipid binding.
Specifications
| C2CD5 | |
| Polyclonal | |
| Unconjugated | |
| C2CD5 | |
| 138 kDa C2 domain-containing phosphoprotein; 5730419I09Rik; C030008B15Rik; C2 calcium dependent domain containing 5; C2 calcium-dependent domain containing 5; C2 domain-containing phosphoprotein of 138 kDa; C2 domain-containing protein 5; C2CD5; Cdp138; KIAA0528; RGD1304592 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 9847 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q86YS7 | |
| C2CD5 | |
| Synthetic peptide directed towards the N-terminal of human C2CD5 (aa 237-286). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction