missing translation for 'onlineSavingsMsg'
Learn More

C2CD3 Antibody [Biotin], Novus Biologicals™

Product Code. 30736250 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30736250 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30736250 Supplier Novus Biologicals Supplier No. AF7348B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

C2CD3 Polyclonal antibody specifically detects C2CD3 in Human samples. It is validated for Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen C2CD3
Applications Immunocytochemistry
Classification Polyclonal
Conjugate Biotin
Formulation PBS
Gene Alias C2 calcium-dependent domain containing 3
Host Species Sheep
Immunogen E. coli-derived recombinant human C2CD3, Met1-Asp109, Accession # Q4AC94
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 26005
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.