missing translation for 'onlineSavingsMsg'
Learn More

C29, MedChemExpress

Product Code. 16483028
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16483028 10 mg 10mg
16459596 50 mg 50mg
16455767 5 mg 5mg
16434336 100 mg 100mg
16430659 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16483028 Supplier Medchem Express Supplier No. HY100461
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress C29 is a Toll-like receptor 2 (TLR2) inhibitor. C29 blocks hTLR2/1 and hTLR2/6 signaling with IC50s of 19.7 and 37.6 μM, respectively.

C29 is a Toll-like receptor 2 (TLR2) inhibitor. C29 blocks hTLR2/1 and hTLR2/6 signaling with IC50s of 19.7 and 37.6 μM, respectively.

Specifications

Chemical Name or Material C29
CAS 363600-92-4
Color Orange
Purity Grade Notes Research
Molecular Formula C16H15NO4
Quantity 10 mg
Solubility Information DMSO : ≥ 30 mg/mL (105.16 mM)
SMILES OC1=C(/C=N/C2=C(C)C(C(O)=O)=CC=C2)C=CC=C1OC
Molecular Weight (g/mol) 285.29
Formula Weight 285.29
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.