missing translation for 'onlineSavingsMsg'
Learn More

C25-140, MedChemExpress

Product Code. 16477717
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16477717 5 mg 5mg
16454988 10 mg 10mg
16415756 100 mg 100mg
16401237 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16477717 Supplier Medchem Express Supplier No. HY120934
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress C25-140, a first-in-class, orally active, and fairly selective TRAF6-Ubc13 inhibitor, directly binds to TRAF6, and blocks the interaction of TRAF6 with Ubc13. C25-140 lowers TRAF6 activity, reduces NF-κB activation, and combats autoimmunity.

C25-140, a first-in-class, orally active, and fairly selective TRAF6-Ubc13 inhibitor, directly binds to TRAF6, and blocks the interaction of TRAF6 with Ubc13. C25-140 lowers TRAF6 activity, reduces NF-κB activation, and combats autoimmunity.

Specifications

Chemical Name or Material C25-140
CAS 1358099-18-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C26H31N7O
Quantity 5 mg
Solubility Information DMSO : 41.67 mg/mL (91.07 mM; Need ultrasonic)
SMILES O=C(N1CCC(CC2=CC=CC=C2)CC1)CCC3=C(C)N(C4=NN5C(C=C4)=NN=C5C)N=C3C
Molecular Weight (g/mol) 457.57
Formula Weight 457.57
Percent Purity 98.12%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.