missing translation for 'onlineSavingsMsg'
Learn More

C24:1 Ceramide, TRC

Product Code. 30179617
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30179617 50 mg 50mg
30178961 10 mg 10mg
30179321 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30179617 Supplier Toronto Research Chemicals Supplier No. TRCC26346050MG
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54164-50-0
Molecular Formula C42 H81 N O3
Molecular Weight (g/mol) 648.1
Synonym (15Z)-N-[(1S,2R,3E)-2-Hydroxy-1-(hydroxymethyl)-3-heptadecen-1-yl]-15-tetracosenamide, 15-Tetracosenamide, N-[(1S,2R,3E)-2-hydroxy-1-(hydroxymethyl)-3-heptadecenyl]-, (15Z)- (9CI), 15-Tetracosenamide, N-[2-hydroxy-1-(hydroxymethyl)-3-heptadecenyl]-, [R-[R*,S*-(E,Z)]]-, (15Z)-N-[(1S,2R,3E)-2-Hydroxy-1-(hydroxymethyl)-3-heptadecen-1-yl]-15-tetracosenamide, C24:1-Ceramide, D-erythro-1,3-Dihydroxy-2-(cis-15-tetracosenoylamido)-trans-4-octadecene, N-Nervonoyl-D-sphingosine, N-cis-15-Tetracosenoyl-C18-sphingosine
IUPAC Name (Z)-N-[(E,2S,3R)-1,3-dihydroxyoctadec-4-en-2-yl]tetracos-15-enamide
SMILES CCCCCCCCCCCCC\C=C\[C@@H](O)[C@H](CO)NC(=O)CCCCCCCCCCCCC\C=C/CCCCCCCC

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C42H81NO3/c1-3-5-7-9-11-13-15-17-18-19-20-21-22-23-24-26-28-30-32-34-36-38-42(46)43-40(39-44)41(45)37-35-33-31-29-27-25-16-14-12-10-8-6-4-2/h17-18,35,37,40-41,44-45H,3-16,19-34,36,38-39H2,1-2H3,(H,43,46)/b18-17-,37-35+/t40-,41+/m0/s1
Formula Weight 647.6216
Chemical Name or Material Nervonyl-D-sphingosine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.