missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C22orf25 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27846-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene belongs to the transport and Golgi organization family, whose members are predicted to play roles in secretory protein loading in the endoplasmic reticulum. Depletion of this gene in Drosophila S2 cells causes fusion of the Golgi with the ER. In mouse tissue culture cells, this protein co-localizes with a mitochondrially targeted mCherry protein and displays very low levels of co-localization with Golgi and peroxisomes. Allelic variants of this gene are associated with rhabdomyolysis, metabolic crises with encephalopathy, and cardiac arrhythmia.Specifications
| C22orf25 | |
| Polyclonal | |
| Unconjugated | |
| TANGO2 | |
| C22orf25, TANGO2, Transport and Golgi organization protein 2 homolog | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 128989, 27883 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P54797, Q6ICL3 | |
| TANGO2 | |
| C22orf25 Fusion Protein Ag27127 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction