missing translation for 'onlineSavingsMsg'
Learn More

C22 Dihydroceramide, TRC

Product Code. 30357715
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30357715 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30357715 Supplier Toronto Research Chemicals Supplier No. TRCC26314550MG
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material C22 Dihydroceramide
CAS 147492-65-7
Molecular Formula C40 H81 N O3
Quantity 50 mg
InChI Formula InChI=1S/C40H81NO3/c1-3-5-7-9-11-13-15-17-18-19-20-21-22-24-26-28-30-32-34-36-40(44)41-38(37-42)39(43)35-33-31-29-27-25-23-16-14-12-10-8-6-4-2/h38-39,42-43H,3-37H2,1-2H3,(H,41,44)/t38-,39+/m0/s1
SMILES CCCCCCCCCCCCCCCCCCCCCC(=O)N[C@@H](CO)[C@H](O)CCCCCCCCCCCCCCC
IUPAC Name N-[(2S,3R)-1,3-dihydroxyoctadecan-2-yl]docosanamide
Molecular Weight (g/mol) 624.08
Formula Weight 623.6216
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.