missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C21orf7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA542201
This item is not returnable.
View return policy
Description
Peptide sequence: DSEESMEVFK QHCQIAEEYH EVKKEITLLE QRKKELIAKL DQAEKEKVDA Sequence homology: Cow: 100%; Dog: 93%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
C21orf7 plays a critical role in the TGF-beta signaling transduction pathway.
Specifications
| C21orf7 | |
| Polyclonal | |
| Unconjugated | |
| MAP3K7CL | |
| C1H21orf7; C21orf7; C31H21orf7; HC21ORF7; MAP3K7 C-terminal like; MAP3K7 C-terminal-like protein; Map3k7cl; ORF63; TAK1L; TAK1-like protein; TAK1-like protein 1; TAK1-like protein 2; TAK1-like protein 4; TAKL; TAKL-1; TAKL-2; TAKL-4; TGF-beta activated kinase; TGF-beta-activated kinase like | |
| Rabbit | |
| Protein A | |
| RUO | |
| 56911 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P57077 | |
| MAP3K7CL | |
| Synthetic peptide directed towards the C-terminal of human C21orf7 (aa 51-100). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction