missing translation for 'onlineSavingsMsg'
Learn More

C21orf59 Antibody, Novus Biologicals™

Product Code. 18425641 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
25 μL
Unit Size:
0.10mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18425641 25 μL 25µL
18474801 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18425641 Supplier Novus Biologicals Supplier No. NBP18827525ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

C21orf59 Polyclonal antibody specifically detects C21orf59 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen C21orf59
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200
Formulation PBS (pH 7.2) and 40% Glycerol
Gene Alias C21orf48, chromosome 21 open reading frame 48, chromosome 21 open reading frame 59, FLJ20467, FLJ37137, FLJ40247, hypothetical protein LOC56683, kur, kurly
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: QVLKKTIEEAKAIISKKQVEAGVCVTMEMVKDALDQLRGAVMIVYPMGLPPYDPIRMEFE
Purification Method Immunogen affinity purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 56683
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.