Learn More
C21orf58 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 25024-1-AP-150UL
 View more versions of this product
Product Code. 16813555
  • £349.00 / 150µL
Specifications
C21orf58
Polyclonal
Unconjugated
C21orf58
C21orf58
Rabbit
Antigen Affinity Chromatography
RUO
54058
-20°C
Liquid
Immunohistochemistry (Paraffin)
0.51 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P58505
C21orf58
C21orf58 Fusion Protein Ag21758
150 μL