missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C20orf43 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA546767
This item is not returnable.
View return policy
Description
Peptide sequence: EKALGKAASH IKSIKNVTEL KLSDNPAWEG DKGNTKGDKH DDLQRARFIC.
C20orf43 protein function remains unknown.
Specifications
| C20orf43 | |
| Polyclonal | |
| Unconjugated | |
| RTF2 | |
| 1700067C04Rik; 2410001C21Rik; 5730427M17Rik; AA589417; AD-007; C20orf43; CDA05; CDAO5; HSPC164; HSPC169; likely orthologue of H. sapiens chromosome 20 open reading frame 43 (C20orf43); protein RTF2 homolog; Replication termination factor 2; replication termination factor 2 domain containing 1; replication termination factor 2 domain-containing protein 1; RGD1311072; Rtf2; RTFDC1; SHUJUN-3; UPF0549 protein C20orf43; UPF0549 protein C20orf43 homolog | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 51507 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9BY42 | |
| RTF2 | |
| Synthetic peptide directed towards the N-terminal of human RTFDC1 (aa 72-121). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction