Learn More
C20orf3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA543374
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: QETVMKFVPR YSLVLELSDS GAFRRSLHDP DGLVATYISE VHEHDGHLYL Sequence homology: Cow: 77%; Dog: 92%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%
C20orf3 Polyclonal antibody specifically detects C20orf3 in Human samples. It is validated for Western Blot.Specifications
C20orf3 | |
Polyclonal | |
Unconjugated | |
APMAP | |
APMAP | |
Synthetic peptide directed towards the C-terminal of human C20orf3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9HDC9 | |
Rabbit | |
Protein A | |
RUO | |
57136 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |