Learn More
Invitrogen™ C20orf3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113899
Description
Immunogen sequence: EPPLLLGVLH PNTKLRQAER LFENQLVGPE SIAHIGDVMF TGTADGRVVK For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%; Zebrafish: 92%.
APMAP (Adipocyte Plasma Membrane Associated Protein) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include arylesterase activity and strictosidine synthase activity. Exhibits strong arylesterase activity with beta-naphthyl acetate and phenyl acetate. May play a role in adipocyte differentiation.
Specifications
| C20orf3 | |
| Polyclonal | |
| Unconjugated | |
| APMAP | |
| Adipocyte plasma membrane-associated protein; APMAP; C20orf3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 57136 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9HDC9 | |
| APMAP | |
| Synthetic peptide directed towards the N terminal region of human C20orf3 (aa 74-123). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.