missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAB5IF Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112195
This item is not returnable.
View return policy
Description
RAB5IF Polyclonal Antibody for ICC/IF, IHC (P)
RAB5IF, also known as RAB5-interacting factor, is a protein that interacts with the small GTPase RAB5 and plays a role in endocytic trafficking. It is involved in the regulation of early endosome fusion and recycling, and has been implicated in various cellular processes such as cell migration and cytokinesis.
Specifications
| RAB5IF | |
| Polyclonal | |
| Unconjugated | |
| C20orf24 | |
| C20orf24; chromosome 20 open reading frame 24; PNAS-11; RAB5IF; Rab5-interacting protein; RIP5; uncharacterized protein C20orf24; Uncharacterized protein RAB5IF | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 55969 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.03% ProClin 300; pH 7 | |
| Q9BUV8 | |
| C20orf24 | |
| E. coli-derived Human C20orf24/RAB5IF/RAB5IF fragment. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction