missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C20orf203 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568728
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Human: 100%.
The protein encoded by this gene is thought to be a human-specific protein. Currently available evidence suggests that orthologous regions in other organisms contain sequence differences that would not support production of a protein product. Genome-wide association studies have suggested the possibility that a SNP in the 3' UTR, rs17123507, could be associated with nicotine addiction. Expression of this gene may be elevated in some individuals with Alzheimer's disease.
Specifications
| C20orf203 | |
| Polyclonal | |
| Unconjugated | |
| C20orf203 | |
| alugen; C20orf203; chromosome 20 open reading frame 203; uncharacterized protein C20orf203 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 284805 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8NBC4 | |
| C20orf203 | |
| synthetic peptide directed towards the C terminal region of human C20orf203. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction