missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C20orf194 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23184-1-AP-20UL
This item is not returnable.
View return policy
Description
The function of the C20orf194 gene has not yet been determined.Specifications
| C20orf194 | |
| Polyclonal | |
| Unconjugated | |
| C20orf194 | |
| C20orf194 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 25943, 499891 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.38 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5TEA3 | |
| C20orf194, RGD1565616 | |
| C20orf194 Fusion Protein Ag19714 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction