missing translation for 'onlineSavingsMsg'
Learn More

C2 Ceramide, TRC

Product Code. 30357681
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30357681 10 mg 10mg
30350942 25 mg 25mg
30355995 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30357681 Supplier Toronto Research Chemicals Supplier No. TRCC26250010MG
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3102-57-6
Molecular Formula C20 H39 N O3
Molecular Weight (g/mol) 341.53
IUPAC Name N-[(E,2R,3S)-1,3-dihydroxyoctadec-4-en-2-yl]acetamide
SMILES CCCCCCCCCCCCC\C=C\[C@H](O)[C@@H](CO)NC(=O)C

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C20H39NO3/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-20(24)19(17-22)21-18(2)23/h15-16,19-20,22,24H,3-14,17H2,1-2H3,(H,21,23)/b16-15+/t19-,20+/m1/s1
Formula Weight 341.293
Chemical Name or Material C2 Ceramide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.